accessibility menu, dialog, popup

UserName

H2OH2OH2ONaNaNaHOONNOOOOOO

CAS RN 150-38-9

IUPAC Name: trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate trihydrate
Synonyms: trisodium edta
Molecular Weight (g/mol): 412.23
Molecular Formula: C10H19N2Na3O11
InChi Key: FXNQQEVEDZAAJM-UHFFFAOYSA-K
SMILES: O.O.O.[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results