accessibility menu, dialog, popup

UserName

OOSOOO

CAS RN 14995-40-5

IUPAC Name: 3-sulfonatobenzoate
Synonyms: 3-sulfonatobenzoate
Molecular Weight (g/mol): 200.17
Molecular Formula: C7H4O5S
InChi Key: QMWGSOMVXSRXQX-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=CC(=C1)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results