accessibility menu, dialog, popup

UserName

CAS RN 1499-55-4

H3COONH2OHO

CAS RN 1499-55-4

IUPAC Name: 2-amino-5-methoxy-5-oxopentanoic acid
Synonyms: 2-amino-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: ZGEYCCHDTIDZAE-UHFFFAOYNA-N
SMILES: COC(=O)CCC(N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)