accessibility menu, dialog, popup

UserName

CAS RN 149849-92-3

NNOOHCl

CAS RN 149849-92-3

IUPAC Name: 2-chloropyrimidine-4-carboxylic acid
Synonyms: 2-chloropyrimidine-4-carboxylic acid
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: YMDSUQSBJRDYLI-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results