CAS RN 149817-62-9
IUPAC Name:
4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms:
4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol):
312.17
Molecular Formula:
C15H10BrN3
InChi Key:
CJRRILXBSCRHKN-UHFFFAOYSA-N
SMILES:
BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
1
–
3
of
3
results
1
4'-Bromo-2,2':6',2' '-terpyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4'-Bromo-2,2':6',2' '-terpyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4'-Bromo-2,2':6',2' '-terpyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.