accessibility menu, dialog, popup

UserName

CH3OPClClO

CAS RN 1498-51-7

IUPAC Name: ethyl chlorophosphonochloridate
Synonyms: ethyl phosphorodichloridate
Molecular Weight (g/mol): 162.93
Molecular Formula: C2H5Cl2O2P
InChi Key: YZBOZNXACBQJHI-UHFFFAOYSA-N
SMILES: CCOP(Cl)(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results