accessibility menu, dialog, popup

UserName

H2NNHOO

CAS RN 149423-77-8

IUPAC Name: benzyl N-(4-aminocyclohexyl)carbamate
Synonyms: benzyl N-(4-aminocyclohexyl)carbamate
Molecular Weight (g/mol): 248.33
Molecular Formula: C14H20N2O2
InChi Key: JQVBZZUMWRXDSQ-UHFFFAOYSA-N
SMILES: NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results