accessibility menu, dialog, popup

UserName

CAS RN 149-64-4

BrH3CNCH3(S)(S)(S)O(R)OOOH

CAS RN 149-64-4

IUPAC Name: (1S,2R,4S,5S)-9-butyl-7-[(3-hydroxy-2-phenylpropanoyl)oxy]-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide
Synonyms: hyoscine butylbromide
Molecular Weight (g/mol): 440.38
Molecular Formula: C21H30BrNO4
InChi Key: HOZOZZFCZRXYEK-GXLSIKBRSA-M
SMILES: [Br-].CCCC[N+]1(C)[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)C(CO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results