accessibility menu, dialog, popup

UserName

CAS RN 148749-58-0

CH3NH2H3CCH3H3C

CAS RN 148749-58-0

IUPAC Name: 1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)methanamine
Synonyms: 1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)methanamine
Molecular Weight (g/mol): 217.36
Molecular Formula: C15H23N
InChi Key: GVSNYOXKXQRGRC-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(CN)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results