accessibility menu, dialog, popup

UserName

CAS RN 14874-82-9

OOH3CH3CCCORhO

CAS RN 14874-82-9

IUPAC Name: 2,2-bis(hydroxymethyl)-4,6-dimethyl-1λ³,3-dioxa-2-rhodacyclohexa-1(6),4-dien-2-yl
Synonyms: 2,2-bis(hydroxymethyl)-4,6-dimethyl-1λ³,3-dioxa-2-rhodacyclohexa-1(6),4-dien-2-yl
Molecular Weight (g/mol): 261.06
Molecular Formula: C7H10O4Rh
InChi Key: APTDRJHXOSPVAT-FGSKAQBVSA-N
SMILES:
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)