accessibility menu, dialog, popup

UserName

CAS RN 148546-99-0

CH3NNNH2

CAS RN 148546-99-0

IUPAC Name: 3-(4-methylpiperazin-1-yl)aniline
Synonyms: 3-(4-methylpiperazin-1-yl)aniline
Molecular Weight (g/mol): 191.28
Molecular Formula: C11H17N3
InChi Key: RJGHJWKQCJAJEP-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC(N)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results