accessibility menu, dialog, popup

UserName

CAS RN 14852-31-4

H3CCH3OH

CAS RN 14852-31-4

IUPAC Name: hexadecan-2-ol
Synonyms: 2-hexadecanol
Molecular Weight (g/mol): 242.45
Molecular Formula: C16H34O
InChi Key: FVDRFBGMOWJEOR-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results