missing translation for 'onlineSavingsMsg'
Learn More

2-Hexadecanol, MP Biomedicals

Product Code. 11411726 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
11411726 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 11411726

Brand: MP Biomedicals 0520755301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14852-31-4
Molecular Formula C16H34O
Quantity 1 g
Synonym 2-hexadecanol, hexadecanol-2, 2-hexadecanol, 2r, 2-hexadecanol, 2s, acmc-1bt5g, acmc-20m2p4, acmc-20m2p5
InChI Key FVDRFBGMOWJEOR-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCC(C)O
IUPAC Name hexadecan-2-ol
Molecular Weight (g/mol) 242.447
PubChem CID 85779
Formula Weight 242.45
Chemical Name or Material 2-Hexadecanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.