accessibility menu, dialog, popup

UserName

CAS RN 1484-84-0

OH(S)H2N

CAS RN 1484-84-0

IUPAC Name: (2S)-2-(2-hydroxyethyl)piperidin-1-ium
Synonyms: (2S)-2-(2-hydroxyethyl)piperidin-1-ium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: PTHDBHDZSMGHKF-ZETCQYMHSA-O
SMILES: OCC[C@@H]1CCCC[NH2+]1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)