accessibility menu, dialog, popup

UserName

CAS RN 1484-13-5

H2CN

CAS RN 1484-13-5

IUPAC Name: 9-ethenyl-9H-carbazole
Synonyms: N-vinylcarbazole
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: KKFHAJHLJHVUDM-UHFFFAOYSA-N
SMILES: C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)