accessibility menu, dialog, popup

UserName

CAS RN 1484-12-4

IUPAC Name: 9-methyl-9H-carbazole
Synonyms: 9H-carbazole, 9-methyl-
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: SDFLTYHTFPTIGX-UHFFFAOYSA-N
SMILES: CN1C2=C(C=CC=C2)C2=C1C=CC=C2
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results