accessibility menu, dialog, popup

UserName

H3COOCH3SH

CAS RN 1483-27-8

IUPAC Name: 2,5-dimethoxybenzene-1-thiol
Synonyms: 2,5-dimethoxybenzenethiol
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: SESUUAOAUZDHHP-UHFFFAOYSA-N
SMILES: COC1=CC(S)=C(OC)C=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)