accessibility menu, dialog, popup

UserName

CAS RN 14815-19-1

NHNOFFF

CAS RN 14815-19-1

IUPAC Name: 2,2,2-trifluoro-N-(pyridin-3-yl)acetamide
Synonyms: 2,2,2-trifluoro-N-(pyridin-3-yl)acetamide
Molecular Weight (g/mol): 190.13
Molecular Formula: C7H5F3N2O
InChi Key: KNGWJNVUCYDZJE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results