accessibility menu, dialog, popup

UserName

CAS RN 148-82-3

H2NNClClHOO

CAS RN 148-82-3

IUPAC Name: 2-amino-3-{4-[bis(2-chloroethyl)amino]phenyl}propanoic acid
Synonyms: alkeran sarcolysine levofolan L-sarkolysin sarkolysin levofalan sarcolysine hydrochloride levopholan mustard, phenylalanine ccris 374
Molecular Weight (g/mol): 305.2
Molecular Formula: C13H18Cl2N2O2
InChi Key: SGDBTWWWUNNDEQ-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)N(CCCl)CCCl)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)