CAS RN 148-24-3
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
CAS: 148-24-3 Molecular Formula: C9H7NO Molecular Weight (g/mol): 145.16 MDL Number: MFCD00006807 InChI Key: MCJGNVYPOGVAJF-UHFFFAOYSA-N Synonym: 8-hydroxyquinoline,8-quinolinol,oxyquinoline,oxine,quinophenol,8-quinol,8-oxyquinoline,phenopyridine,oxychinolin,bioquin PubChem CID: 1923 ChEBI: CHEBI:48981 IUPAC Name: quinolin-8-ol SMILES: OC1=C2N=CC=CC2=CC=C1
8-Hydroxyquinoline, ACS
CAS: 148-24-3 Molecular Formula: C9H7NO Molecular Weight (g/mol): 145.16 MDL Number: MFCD00006807 InChI Key: MCJGNVYPOGVAJF-UHFFFAOYSA-N Synonym: 8-hydroxyquinoline,8-quinolinol,oxyquinoline,oxine,quinophenol,8-quinol,8-oxyquinoline,phenopyridine,oxychinolin,bioquin PubChem CID: 1923 ChEBI: CHEBI:48981 IUPAC Name: quinolin-8-ol SMILES: OC1=C2N=CC=CC2=CC=C1
8-Hydroxyquinoline, 99%
CAS: 148-24-3 Molecular Formula: C9H7NO Molecular Weight (g/mol): 145.16 MDL Number: MFCD00006807 InChI Key: MCJGNVYPOGVAJF-UHFFFAOYSA-N Synonym: 8-hydroxyquinoline,8-quinolinol,oxyquinoline,oxine,quinophenol,8-quinol,8-oxyquinoline,phenopyridine,oxychinolin,bioquin PubChem CID: 1923 ChEBI: CHEBI:48981 IUPAC Name: quinolin-8-ol SMILES: OC1=C2N=CC=CC2=CC=C1
Oxyquinoline (8-Hydroxyquinoline), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Oxyquinoline (8-Hydroxyquinoline), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
8-Hydroxyquinoline, Sigma-Aldrich™
CAS: 148-24-3 Grade: Research Molecular Weight (g/mol): 145.16
8-Hydroxyquinoline, TRC
CAS: 148-24-3 Molecular Formula: C9 H7 N O Molecular Weight (g/mol): 145.16 Synonym: 8-Quinolinol,1-Azanaphthalene-8-ol,8-Hydroxychinolin,8-Hydroxyquinoline,8-OQ,8-Oxyquinoline,8-Quinol,AQ+,Albisal,Fennosan H 30,NSC 2039,NSC 285166,NSC 402623,NSC 48037,NSC 54230,NSC 615011,NSC 82404,NSC 82405,NSC 82409,NSC 82410,NSC 82412,Oxin,Oxine,Oxoquinoline,Oxychinolin,Oxyquinoline,Phenopyridine,Quinophenol,Tumex IUPAC Name: quinolin-8-ol SMILES: Oc1cccc2cccnc12
8-Hydroxyquinoline, MedChemExpress
MedChemExpress 8-Hydroxyquinoline (8-Hydroxyquinoline) is a monoprotic bidentate chelating agent, exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.