accessibility menu, dialog, popup

UserName

CAS RN 14799-82-7

BrP

CAS RN 14799-82-7

IUPAC Name: (cyclopropylmethyl)triphenylphosphanium bromide
Synonyms: (cyclopropylmethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 397.3
Molecular Formula: C22H22BrP
InChi Key: WFQSHRSBITUSIB-UHFFFAOYSA-M
SMILES: [Br-].C(C1CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results