accessibility menu, dialog, popup

UserName

CH3CH3CH3OONO

CAS RN 147804-30-6

IUPAC Name: tert-butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Synonyms: tert-butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Molecular Weight (g/mol): 213.28
Molecular Formula: C11H19NO3
InChi Key: ULSBMKGFFFMGOI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2(CO2)CC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results