accessibility menu, dialog, popup

UserName

CAS RN 1478-61-1

IUPAC Name: 4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
Synonyms: bisphenol AF hexafluoroacetone bisphenol A biphenol AF hexafluorodiphenylolpropane
Molecular Weight (g/mol): 336.23
Molecular Formula: C15H10F6O2
InChi Key: ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results