accessibility menu, dialog, popup

UserName

CAS RN 14774-37-9

IUPAC Name: (oxan-4-yl)methanol
Synonyms: oxan-4-ylmethanol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: YSNVSVCWTBLLRW-UHFFFAOYSA-N
SMILES: OCC1CCOCC1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)