CAS RN 1477-42-5
CAS RN 1477-42-5
IUPAC Name:
4-methyl-1,3-benzothiazol-2-amine
Synonyms:
2-benzothiazolamine, 4-methyl-
Molecular Weight (g/mol):
164.23
Molecular Formula:
C8H8N2S
InChi Key:
GRIATXVEXOFBGO-UHFFFAOYSA-N
SMILES:
CC1=C2N=C(N)SC2=CC=C1
1
–
1
of
1
results
1
2-Amino-4-methylbenzothiazole, 98%
CAS: 1477-42-5 Molecular Formula: C8H8N2S Molecular Weight (g/mol): 164.23 MDL Number: MFCD00005793 InChI Key: GRIATXVEXOFBGO-UHFFFAOYSA-N Synonym: 2-amino-4-methylbenzothiazole,4-methylbenzo d thiazol-2-amine,2-benzothiazolamine, 4-methyl,4-methyl-2-aminobenzothiazole,4-methyl-2-benzothiazolamine,benzothiazole, 2-amino-4-methyl,4-methylbenzothiazol-2-ylamine,2-amino-4-methyl benzothiazole,4-methyl-2-aminobenzothiozole,unii-45v5f1b9z8 PubChem CID: 15132 IUPAC Name: 4-methyl-1,3-benzothiazol-2-amine SMILES: CC1=C2N=C(N)SC2=CC=C1