accessibility menu, dialog, popup

UserName

CAS RN 14731-14-7

BrOON

CAS RN 14731-14-7

IUPAC Name: 2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethan-1-one
Synonyms: 2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
Molecular Weight (g/mol): 266.09
Molecular Formula: C11H8BrNO2
InChi Key: XTBXGZOVSCTNEC-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results