CAS RN 1471-56-3
CAS RN 1471-56-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Decarboxamide Meprobamate, TRC
CAS: 1471-56-3 Molecular Formula: C8 H17 N O3 Molecular Weight (g/mol): 175.23 Synonym: 2-Methyl-2-propyl-1,3-propanediol Monocarbamate,Carbamic Acid 2-(Hydroxymethyl)-2-methylpentyl Ester,NSC 108719, Carisoprodol Related Compound A IUPAC Name: [2-(hydroxymethyl)-2-methylpentyl] carbamate SMILES: CCCC(C)(CO)COC(=O)N