accessibility menu, dialog, popup

UserName

CAS RN 147-93-3

HOOHS

CAS RN 147-93-3

IUPAC Name: 2-sulfanylbenzoic acid
Synonyms: thiosalicylic acid o-mercaptobenzoesaeure o-benzoic acid thiol
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: NBOMNTLFRHMDEZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1S
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)