accessibility menu, dialog, popup

UserName

CAS RN 146954-74-7

(R)OO(R)N(R)HNO(R)HOOOOH3CH3CF

CAS RN 146954-74-7

IUPAC Name: 1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl)-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 548.57
Molecular Formula: C30H29FN2O7
InChi Key: CSSFZSSZXOCCJB-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(OCC1OC(C(F)C1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results