accessibility menu, dialog, popup

UserName

CH3ONHH3CH3CNOH3CCH3

CAS RN 14691-89-5

IUPAC Name: (4-acetamido-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: 4-acetamido-2,2,6,6-tetramethylpiperidin-1-yloxidanyl
Molecular Weight (g/mol): 213.3
Molecular Formula: C11H21N2O2
InChi Key: UXBLSWOMIHTQPH-UHFFFAOYSA-N
SMILES: CC(=O)NC1CC(C)(C)N([O])C(C)(C)C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results