accessibility menu, dialog, popup

UserName

CAS RN 1469-94-9

IUPAC Name: 1-(2-hydroxyphenyl)-3-phenylpropane-1,3-dione
Synonyms: 1-(2-hydroxyphenyl)-3-phenylpropane-1,3-dione
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: OABFIJGAEVKMJP-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)CC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results