CAS RN 14676-01-8
IUPAC Name:
(3R)-3-(benzylazaniumyl)butanoate
Synonyms:
(3R)-3-(benzylammonio)butanoate
Molecular Weight (g/mol):
193.25
Molecular Formula:
C11H15NO2
InChi Key:
QIXZNMWEYBPTEQ-SECBINFHSA-N
SMILES:
C[C@H](CC([O-])=O)[NH2+]CC1=CC=CC=C1