accessibility menu, dialog, popup

UserName

CAS RN 14675-44-6

CH3NNOHCH3

CAS RN 14675-44-6

IUPAC Name: (1,4-dimethylpiperazin-2-yl)methanol
Synonyms: (1,4-dimethylpiperazin-2-yl)methanol
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: ZEHZRJZZIYNPKU-UHFFFAOYNA-N
SMILES: CN1CCN(C)C(CO)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results