CAS RN 1466-82-6
CAS RN 1466-82-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Acetylsalicylic Acid EP Impurity F (Carbasalate Calcium EP Impurity A), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Acetylsalicylic Anhydride, TRC
CAS: 1466-82-6 Molecular Formula: C18 H14 O7 Molecular Weight (g/mol): 342.3 Synonym: 2-(Acetyloxy)benzoic Anhydride,Carbasalate Calcium Imp. A (EP),Acetylsalicylic Acid Imp. F (EP),Acetylsalicylic Anhydride IUPAC Name: (2-acetyloxybenzoyl) 2-acetyloxybenzoate SMILES: CC(=O)Oc1ccccc1C(=O)OC(=O)c2ccccc2OC(=O)C