accessibility menu, dialog, popup

UserName

CAS RN 146575-64-6

CH3OOFFFOH

CAS RN 146575-64-6

IUPAC Name: 1-[2-hydroxy-5-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[2-hydroxy-5-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: FYWYHQOZXGAPSF-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=CC(OC(F)(F)F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results