accessibility menu, dialog, popup

UserName

CAS RN 14649-73-1

NaNaNaCoOHNOOHNOOHNOOHNOONHOONHO

CAS RN 14649-73-1

IUPAC Name: cobalt(3+) trisodium hexakis(dioxoamine)
Synonyms: cobalt(3+) trisodium hexakis(nitrite ion)
Molecular Weight (g/mol): 409.98
Molecular Formula: CoH6N6Na3O12
InChi Key: QLNJFTGEDYRKNP-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[Co+3].[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)