accessibility menu, dialog, popup

UserName

CAS RN 14640-21-2

MgNNNN

CAS RN 14640-21-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 637.04
Molecular Formula: C44H28MgN4
InChi Key: XEHJAWQTIIXDON-QHNUXLDVSA-N
SMILES: C1=CC2=C(C3=[N]4C(C=C3)=C(C3=CC=C5N3[Mg]43N2C1=C(C1=[N]3C(C=C1)=C5C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results