accessibility menu, dialog, popup

UserName

OO

CAS RN 1464-53-5

IUPAC Name: 2,2'-bioxirane
Synonyms: diepoxybutane bioxirane dioxybutadiene butane diepoxide 1,2:3,4-diepoxybutane DL butadiene dioxide 1,1'-bi[ethylene oxide] 1,3-butadiene diepoxide 1,2:3,4-butadiene diepoxide butadiendioxyd
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: ZFIVKAOQEXOYFY-UHFFFAOYNA-N
SMILES: C1OC1C1CO1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)