accessibility menu, dialog, popup

UserName

H3COFO

CAS RN 146137-74-8

IUPAC Name: 2-fluoro-6-methoxybenzaldehyde
Synonyms: 2-fluoro-6-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: UIOAYOIJMYMOEU-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C(F)=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results