accessibility menu, dialog, popup

UserName

PdClClNNH3CCH3

CAS RN 14592-56-4

IUPAC Name: palladium(2+) bis(acetonitrile) dichloride
Synonyms: palladium(2+) bis(acetonitrile) dichloride
Molecular Weight (g/mol): 259.43
Molecular Formula: C4H6Cl2N2Pd
InChi Key: RBYGDVHOECIAFC-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].CC#N.CC#N
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)