accessibility menu, dialog, popup

UserName

CAS RN 14589-44-7

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHTm

CAS RN 14589-44-7

IUPAC Name: tris(4-hydroxypent-3-en-2-one) thulium
Synonyms: tris(4-hydroxypent-3-en-2-one) thulium
Molecular Weight (g/mol): 469.29
Molecular Formula: C15H24O6Tm
InChi Key: BKQDCXRDXKJCRG-UHFFFAOYSA-N
SMILES: [Tm].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results