accessibility menu, dialog, popup

UserName

CAS RN 1455-21-6

H3CSH

CAS RN 1455-21-6

IUPAC Name: nonane-1-thiol
Synonyms: 1-nonanethiol
Molecular Weight (g/mol): 160.32
Molecular Formula: C9H20S
InChi Key: ZVEZMVFBMOOHAT-UHFFFAOYSA-N
SMILES: CCCCCCCCCS
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)