accessibility menu, dialog, popup

UserName

CAS RN 145428-94-0

CH3ON(S)(R)(S)OOHN(S)(S)(R)OCH3

CAS RN 145428-94-0

IUPAC Name: (1S,11S,18S,20R,21R,22S)-4,5-dimethoxy-9-oxo-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-17-ium
Synonyms: (1S,11S,18S,20R,21R,22S)-4,5-dimethoxy-9-oxo-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-17-ium
Molecular Weight (g/mol): 395.48
Molecular Formula: C23H27N2O4
InChi Key: RRKTZKIUPZVBMF-IBTVXLQLSA-O
SMILES: COC1=CC2=C(C=C1OC)[C@@]13CC[NH+]4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results