accessibility menu, dialog, popup

UserName

CAS RN 14523-22-9

CCRhOOClCCRhOOCl

CAS RN 14523-22-9

IUPAC Name: bis([bis(hydroxymethyl)rhodio]-λ²-chlorane)
Synonyms: bis([bis(hydroxymethyl)rhodio]-λ²-chlorane)
Molecular Weight (g/mol): 388.75
Molecular Formula: C4Cl2O4Rh2
InChi Key: FGKDXBICTVUSPK-UHFFFAOYSA-L
SMILES: [Cl-][Rh+](C#[O])C#[O].[Cl-][Rh+](C#[O])C#[O]
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)