accessibility menu, dialog, popup

UserName

CAS RN 1451391-89-1

BOHNCH3ClHOOH

CAS RN 1451391-89-1

IUPAC Name: [2-chloro-4-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 2-chloro-4-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 213.42
Molecular Formula: C8H9BClNO3
InChi Key: LSMHGCJWZPHACP-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(Cl)=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results