CAS RN 144978-82-5
CAS RN 144978-82-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tyrphostin AG 112, TRC
CAS: 144978-82-5 Molecular Formula: C13H8N4O Molecular Weight (g/mol): 236.23 IUPAC Name: (3Z)-2-amino-4-(4-hydroxyphenyl)buta-1,3-diene-1,1,3-tricarbonitrile SMILES: NC(=C(C#N)C#N)\C(=C\c1ccc(O)cc1)\C#N