accessibility menu, dialog, popup

UserName

SFFFFFClOOH

CAS RN 1448317-67-6

IUPAC Name: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 282.61
Molecular Formula: C7H4ClF5O2S
InChi Key: QEUCPMZEYUIRBW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results