accessibility menu, dialog, popup

UserName

CAS RN 144808-72-0

OOI

CAS RN 144808-72-0

IUPAC Name: 4-(4-iodophenoxy)oxane
Synonyms: 4-(4-iodophenoxy)oxane
Molecular Weight (g/mol): 304.13
Molecular Formula: C11H13IO2
InChi Key: PQUFKWFIWPZFBM-UHFFFAOYSA-N
SMILES: IC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results