accessibility menu, dialog, popup

UserName

CAS RN 14464-29-0

CH3OONOO

CAS RN 14464-29-0

IUPAC Name: 2,5-dioxopyrrolidin-1-yl acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl acetate
Molecular Weight (g/mol): 157.13
Molecular Formula: C6H7NO4
InChi Key: SIFCHNIAAPMMKG-UHFFFAOYSA-N
SMILES: CC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)