accessibility menu, dialog, popup

UserName

CAS RN 14456-34-9

IUPAC Name: hafnium nonahydrate dichloride
Synonyms: hafnium nonahydrate dichloride
Molecular Weight (g/mol): 411.53
Molecular Formula: Cl2H18HfO9
InChi Key: CWGPOJHZSBOIPU-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf]
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results